missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Acetyl beta-methylcholine bromide, 98%

Product Code. 11347016
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11347016 5 g 5g
11357016 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11347016 Supplier Alfa Aesar™ Supplier No. L06848.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 333-31-3
Molecular Formula C8H18BrNO2
Molecular Weight (g/mol) 240.14
MDL Number MFCD00011816
InChI Key MMVPLEUBMWUYIB-UHFFFAOYNA-M
Synonym methacholine bromide, 2-acetoxy-n,n,n-trimethylpropan-1-aminium bromide, acetyl beta-methylcholine bromide, 1-propanaminium, 2-acetyloxy-n,n,n-trimethyl-, bromide, 1-propanaminium, 2-acetyloxy-n,n,n-trimethyl-, bromide 1:1, methacholini bromidum, methacholine bromide nf, c8h18no2.br, mecholyl bromide, methacholinebromide
PubChem CID 92754
IUPAC Name [2-(acetyloxy)propyl]trimethylazanium bromide
SMILES [Br-].CC(C[N+](C)(C)C)OC(C)=O

Specifications

Melting Point 147°C to 151°C
Quantity 5 g
Beilstein 6098010
Formula Weight 240.15
Percent Purity 98%
Chemical Name or Material Acetyl beta-methylcholine bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.