missing translation for 'onlineSavingsMsg'
Learn More

Acetyl-beta-methylcholine chloride, 98%, ACROS Organics™

Product Code. 10071214 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10071214 25 g Glass bottle 25g
10031264 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10071214 Supplier Thermo Scientific Acros Supplier No. 102610250

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 62-51-1
Molecular Formula C8H18ClNO2
Molecular Weight (g/mol) 195.687
MDL Number MFCD00011817
InChI Key JHPHVAVFUYTVCL-UHFFFAOYSA-M
Synonym methacholine chloride, acetyl-beta-methylcholine chloride, provocholine, mecholyl chloride, amechol, mecholine chloride, 1-propanaminium, 2-acetyloxy-n,n,n-trimethyl-, chloride, methacholinium chloride, 2-acetoxypropyl trimethylammonium chloride
PubChem CID 6114
ChEBI CHEBI:50142
IUPAC Name 2-acetyloxypropyl(trimethyl)azanium;chloride
SMILES CC(C[N+](C)(C)C)OC(=O)C.[Cl-]

Specifications

CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula CH3CO2CH(CH3)CH2N(CH3)3Cl
Quantity 25 g
Merck Index 15, 6012
Formula Weight 195.69
Chemical Name or Material Acetyl-β-methylcholine chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.