missing translation for 'onlineSavingsMsg'
Learn More

Acetylcholinesterase/ACHE Antibody, Novus Biologicals™

Goat Polyclonal Antibody

Brand:  Novus Biologicals NB100-1519

Product Code. 18048873

  • 531.00€ / 0.10mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Acetylcholinesterase/ACHE Polyclonal specifically detects Acetylcholinesterase/ACHE in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Peptide ELISA, Immunohistochemistry Free-Floating.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Acetylcholinesterase/ACHE
Polyclonal
Unconjugated
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
acetylcholinesterase, acetylcholinesterase (Yt blood group), AChE, apoptosis-related acetylcholinesterase, ARACHE, EC 3.1.1, EC 3.1.1.7, N-ACHE, YT, Yt blood group
Goat
Affinity Purified
RUO
Primary
This antibody is expected to recognise isoform NP_000656.1 only (the ubiquitously expressed, hydrophillic form).
Store at -20C. Avoid freeze-thaw cycles.
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, ELISA
0.5 mg/mL
Western Blot 0.3 - 1 μg/mL, Flow Cytometry 10 μg/mL, Immunocytochemistry/Immunofluorescence 10 μg/mL, Peptide ELISA Detection limit 1:32000, Immunohistochemistry Free-Floating
NP_000656.
ACHE
Peptide with sequence QFDHYSKQDRCSDL corresponding to C-Terminus according to NP_000656.1.
0.1 mg
Cellular Markers
43
Human, Rat
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only