missing translation for 'onlineSavingsMsg'
Learn More

Acetylsalicylic Acid EP Impurity F (Carbasalate Calcium EP Impurity A), Mikromol™

Product Code. 30052614
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30052614 100 mg 100mg
30051191 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30052614 Supplier Mikromol Supplier No. MM0133.02
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Acetylsalicylic Acid EP Impurity F (Carbasalate Calcium EP Impurity A)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-(Acetyloxy)benzoic Anhydride (Acetylsalicylic Anhydride)
CAS 1466-82-6
Analyte or Component Names 2-(Acetyloxy)benzoic Anhydride (Acetylsalicylic Anhydride)
Shipping Condition Ice pack (-20°C)
Molecular Formula C18 H14 O7
Certifications/Compliance ISO 9001
Synonym Acetylsalicylic Acid Imp. F (EP), Carbasalate Calcium Imp. A (EP), Carbasalate Imp. A (EP), 2-(Acetyloxy)benzoic Anhydride, Acetylsalicylic Anhydride, Acetylsalicylic Acid Impurity F, Carbasalate Calcium Impurity A
InChI Formula InChI=1S/C18H14O7/c1-11(19)23-15-9-5-3-7-13(15)17(21)25-18(22)14-8-4-6-10-16(14)24-12(2)20/h3-10H,1-2H3
SMILES CC(=O)Oc1ccccc1C(=O)OC(=O)c2ccccc2OC(=O)C
IUPAC Name (2-acetyloxybenzoyl) 2-acetyloxybenzoate
Molecular Weight (g/mol) 342.3
Quantity 100 mg
Formula Weight 342.074 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.