missing translation for 'onlineSavingsMsg'
Learn More

Acetylsalicylic Anhydride, TRC

Product Code. 30133262
Change view
Click to view available options
Quantity:
10 mg
25 mg
250 mg
Unit Size:
10mg
250mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133262 250 mg 250mg
30011125 10 mg 10mg
30010836 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133262 Supplier Toronto Research Chemicals Supplier No. TRCA187785250MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1466-82-6
Molecular Formula C18 H14 O7
Molecular Weight (g/mol) 342.3
Synonym 2-(Acetyloxy)benzoic Anhydride, Carbasalate Calcium Imp. A (EP), Acetylsalicylic Acid Imp. F (EP), Acetylsalicylic Anhydride
IUPAC Name (2-acetyloxybenzoyl) 2-acetyloxybenzoate
SMILES CC(=O)Oc1ccccc1C(=O)OC(=O)c2ccccc2OC(=O)C

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C18H14O7/c1-11(19)23-15-9-5-3-7-13(15)17(21)25-18(22)14-8-4-6-10-16(14)24-12(2)20/h3-10H,1-2H3
Formula Weight 342.074
Percent Purity >95%
Chemical Name or Material 2-(Acetyloxy)benzoic Anhydride (Acetylsalicylic Anhydride)
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.