missing translation for 'onlineSavingsMsg'
Learn More

Acid Green 5 MP Biomedicals

Product Code. 11477982 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11477982 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11477982 Supplier MP Biomedicals Supplier No. 0215264125
 Restricted Item
Estimated Shipment: 22-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Histological and cytological stain used for plasma counterstaining in laboratory procedures.

Specifications

CAS 5141-20-8
Molecular Formula C37H34N2Na2O9S3
Synonym Acid Green 5, FD & C Green No 2
Color Green
Quantity 25 g
InChI Key DGOBMKYRQHEFGQ-UHFFFAOYSA-L
SMILES CCN(CC1=CC(=CC=C1)S(=O)(=O)[O-])C2=CC=C(C=C2)C(=C3C=CC(=[N+](CC)CC4=CC(=CC=C4)S(=O)(=O)[O-])C=C3)C5=CC=C(C=C5)S(=O)(=O)[O-].[Na+].[Na+]
IUPAC Name disodium;3-[[N-ethyl-4-[[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]cyclohexa-2,5-dien-1-ylidene]-(4-sulfonatophenyl)methyl]anilino]methyl]benzenesulfonate
Molecular Weight (g/mol) 792.844
PubChem CID 21223
ChEBI CHEBI:87065
Formula Weight 792.8
Grade Histological
Chemical Name or Material Acid Green 5
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.