Learn More
Acid Phosphotungstic, Fisher Chemical™
BIOPHARMA
Product Code. 12930494 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12930494 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12930494 Supplier Fisher Chemical Supplier No. RP 156 100
  • ONLINE EXCLUSIVE PRICE
    98.60€
    89.32€ / 100mL
    Save 9.28€
    9% Off
 Restricted Item
Estimated Shipment: 22-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Pharmacopoeia reagents are intended for use in pharmacopoeia testing and monographs, produced under a GMP compliant ISO 9001 quality management system according to the relevant pharmacopoeia specification, using the pharmacopoeia reagent quality components and water meeting the relevant pharmacopoeia monograph requirement for Purified Water. Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO Guide 43.
Specifications
Quantity 100 mL
compliance-icons compliance-icons
Product Identifier
  • Phosphotungstic acid solution. European Pharmacopoeia Ref: 1065200
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H290-May be corrosive to metals.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash hands, forarms and face thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P390-Absorb spillage to prevent material damage.
Supplemental information
  • MIXTURE LIST-Contain: sodium tungstate, dihydrate, phosphoricacid