Learn More
Abcam Acidic Mammalian Chitinase Activity Kit (Fluorometric)
Product Code. 32113690 Shop All Abcam Products
Change view
Click to view available options
Quantity:
100 Tests
Unit Size:
100 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32113690 100 Tests 100 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 32113690 Supplier Abcam Supplier No. ab273291100Test
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Acidic Mammalian Chitinase Activity Kit (Fluorometric) (ab273291) utilizes the ability of AMCase to cleave a synthetic substrate to release the free fluorophore which can be easily quantified (Ex/Em= 320/445 nm) and it uses a specific Lysis Buffer which distinguishes AMCase activity from CHIT1 ac...
Acidic Mammalian Chitinase Activity Kit (Fluorometric) (ab273291) utilizes the ability of AMCase to cleave a synthetic substrate to release the free fluorophore which can be easily quantified (Ex/Em= 320/445 nm) and it uses a specific Lysis Buffer which distinguishes AMCase activity from CHIT1 activity.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Quantity 100 Tests