missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AcmNPV gp64 Protein Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-12764-50ul
This item is not returnable.
View return policy
Description
AcmNPV gp64 Protein Polyclonal antibody specifically detects AcmNPV gp64 Protein in Virus samples. It is validated for Western Blot, ELISA
Specifications
| AcmNPV gp64 Protein | |
| Polyclonal | |
| Western Blot 1:1000-1:5000, ELISA 1:5000-1:10000 | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Virus | |
| Purified |
| Western Blot, ELISA | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Produced in rabbits immunized with purified, recombinant AcmNPV gp64 Protein (Accession#: NP_054158.1; Met1-Thr481). | |
| 50 μg | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction