missing translation for 'onlineSavingsMsg'
Learn More

Acriflavine Hydrochloride (>90%), TRC

Product Code. 30138734
Change view
Click to view available options
Quantity:
100 mg
1 g
10 g
Unit Size:
100mg
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138734 1 g 1g
30132536 10 g 10g
30007740 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138734 Supplier Toronto Research Chemicals Supplier No. TRCA1910901G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69235-50-3
Molecular Formula C27H27Cl3N6
Molecular Weight (g/mol) 541.9
Synonym Acriflavine Dihydrochloride, Acriflavinium Chloride, Flavacridine Hydrochloride, NSC 689003
IUPAC Name acridine-3,6-diamine;10-methylacridin-10-ium-3,6-diamine;chloride;dihydrochloride
SMILES NC1=CC2=C(C=C1)C=C3C(C=C(N)C=C3)=[N+]2C.NC4=CC5=C(C=C4)C=C6C(C=C(N)C=C6)=N5.[Cl-]

Specifications

Quantity 1 g
InChI Formula InChI=1S/C14H13N3.C13H11N3.3ClH/c1-17-13-7-11(15)4-2-9(13)6-10-3-5-12(16)8-14(10)17;14-10-3-1-8-5-9-2-4-11(15)7-13(9)16-12(8)6-10;;;/h2-8H,1H3,(H3,15,16);1-7H,14-15H2;3*1H
Formula Weight 540.1363
Chemical Name or Material Acriflavine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.