missing translation for 'onlineSavingsMsg'
Learn More

Acrivastine, TRC

Product Code. 30132911
Change view
Click to view available options
Quantity:
10 mg
50 mg
250 mg
Unit Size:
10mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30132911 250 mg 250mg
30005383 50 mg 50mg
17977174 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132911 Supplier Toronto Research Chemicals Supplier No. TRCA191180250MG
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87848-99-5
Molecular Formula C22 H24 N2 O2
Molecular Weight (g/mol) 348.44
Synonym 2-Propenoic acid, 3-[6-[(1E)-1-(4-methylphenyl)-3-(1-pyrrolidinyl)-1-propen-1-yl]-2-pyridinyl]-, (2E)-, 2-Propenoic acid, 3-[6-[(1E)-1-(4-methylphenyl)-3-(1-pyrrolidinyl)-1-propenyl]-2-pyridinyl]-, (2E)- (9CI), 2-Propenoic acid, 3-[6-[1-(4-methylphenyl)-3-(1-pyrrolidinyl)-1-propenyl]-2-pyridinyl]-, (E,E)-, Acrivastine, BW 0270C, BW 825C, BW A825C, Semprex, Acrivastine
IUPAC Name (E)-3-[6-[(E)-1-(4-methylphenyl)-3-pyrrolidin-1-ylprop-1-enyl]pyridin-2-yl]prop-2-enoic acid
SMILES Cc1ccc(cc1)\C(=C/CN2CCCC2)\c3cccc(\C=C\C(=O)O)n3

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C22H24N2O2/c1-17-7-9-18(10-8-17)20(13-16-24-14-2-3-15-24)21-6-4-5-19(23-21)11-12-22(25)26/h4-13H,2-3,14-16H2,1H3,(H,25,26)/b12-11+,20-13+
Formula Weight 348.1838
Chemical Name or Material Acrivastine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.