missing translation for 'onlineSavingsMsg'
Learn More

Acrylamide-13C, TRC

Product Code. 18012430
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18012430 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18012430 Supplier Toronto Research Chemicals Supplier No. TRCA19133310MG
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 287399-24-0
Molecular Formula 13CC2H5NO
Quantity 10 mg
Synonym 2-Propenamide-1-13C (9CI), Acrylamide-1-13C, Acrylamide-13C
InChI Formula InChI=1S/C3H5NO/c1-2-3(4)5/h2H,1H2,(H2,4,5)/i3+1
SMILES N[13C](=O)C=C
IUPAC Name (1^{13}C)prop-2-enamide
Molecular Weight (g/mol) 72.07
Formula Weight 72.0405
Chemical Name or Material Acrylamide-13C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.