Learn More
Brady™ Acrylic Retro Reflective Sheeting Marking Tape
Product Code. 11771925 Shop All Brady Products
Change view
Click to view available options
Color:
Black and White Striped
Black and Yellow Striped
Red on White
Dimensions (L x W):
4.57 m x 50.80 mm
4.57 m x 76.20 mm
Unit Size:
1 roll
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Dimensions (L x W) Unit Size
11771925 Black and White Striped 4.57 m x 50.80 mm 1 roll
11761925 Black and Yellow Striped 4.57 m x 50.80 mm 1 roll
11781925 Black and Yellow Striped 4.57 m x 76.20 mm 1 roll
11701645 Red on White 4.57 m x 50.80 mm 1 roll
11711645 Red on White 4.57 m x 76.20 mm 1 roll
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 11771925 Supplier Brady™ Supplier No. 076310
  • ONLINE EXCLUSIVE PRICE
    37.90€
    34.23€ / 1 rolls
    Save 3.67€
    9% Off
Estimated Shipment: 08-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The marking of transport routes must be both flexible and durable, as they are constantly adapted to the changing demands and needs within your facility or warehouse. This Acrylic Retro Reflective Sheeting Marking Tape gives an extra margin of safety during blackouts, at night, or in low-light conditions.
  • The marking of transport routes must be both flexible and durable, as they are constantly adapted to the changing demands and needs within your facility or warehouse.
  • These tapes give an extra margin of safety during blackouts, at night, or in low-light conditions.
Specifications
Alternate Part Number B957 2 in. X5YDS B/W STRIPED REFLECTIVE TAPE
Certifications/Compliance ASTM D 2979, ASTM D 523
Dimensions (L x W) 4.57 m x 50.80 mm
Finish Semi-gloss
For Use With (Application) Utility plant markings, Reflective Bradystake Warning Stakes, Reflective warning stripes
Length (English) 15 ft.
Length (Metric) 4.57 m
Packaging Quantity 1 Roll
Temperature Range (English) -40°F to +158°F
Temperature Range (Metric) -40°C to +70°C
Thickness 0.081 mm
Width (English) 2 in.
Width (Metric) 50.8 mm
Adhesive Type Permanent Acrylic
Color Black and White Striped
Material Plastic
Product Type Reflective Stripes Tape