missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACSF3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25484-1-AP-20UL
This item is not returnable.
View return policy
Specifications
| ACSF3 | |
| Polyclonal | |
| Unconjugated | |
| ACSF3 | |
| ACSF3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 197322, 257633 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.43 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q3URE1, Q4G176 | |
| ACSF3 | |
| ACSF3 Fusion Protein Ag21979 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu