missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ACSL1 Recombinant Rabbit Monoclonal Antibody (7G0L10)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA558014

 View more versions of this product

Product Code. 30283204

  • 457.00€ / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: ATRPKPLKPP CDLSMQSVEV AGSGGARRSA LLDSDEPLVY FYDDVTTLYE GFQRGIQVSN NGPCLGSRKP DQPYEWLSYK QVAELSECIG SALIQKGFKT.

The protein encoded by this gene is an isozyme of the long-chain fatty-acid-coenzyme A ligase family. Although differing in substrate specificity, subcellular localization, and tissue distribution, all isozymes of this family convert free long-chain fatty acids into fatty acyl-CoA esters, and thereby play a key role in lipid biosynthesis and fatty acid degradation.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ACSL1
Recombinant Monoclonal
1.49 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3
P18163, P41216
ACSL1
A synthetic peptide corresponding to a sequence within amino acids 45-144 of human ACSL1 (NP_001986.2).
100 μL
Primary
Mouse, Rat
Antibody
IgG
ELISA, Western Blot
7G0L10
Unconjugated
ACSL1
Acas; Acas1; ACS; Acs1; ACSL1; Acsl2; Acyl CoA synthetase, long chain; acyl-CoA synthetase 1; acyl-CoA synthetase long chain family member 1; acyl-CoA synthetase long-chain family member 1; Arachidonate--CoA ligase; COAA; FACL1; Facl2; FACS; fatty acid Coenzyme A ligase, long chain 2; fatty-acid-Coenzyme A ligase long chain 2; fatty-acid-Coenzyme A ligase, long-chain 1; fatty-acid-Coenzyme A ligase, long-chain 2; L LACS2; LACS; LACS 1; LACS 2; LACS1; LACS2; lignoceroyl-CoA synthase; long-chain acyl-CoA synthetase 1; Long-chain acyl-CoA synthetase 2; long-chain fatty acid-CoA ligase 2; long-chain fatty-acid-coenzyme A ligase 1; long-chain-fatty-acid--CoA ligase 1; Long-chain-fatty-acid--CoA ligase, liver isozyme; OTTHUMP00000218457; OTTHUMP00000218460; OTTHUMP00000218462; OTTHUMP00000218465; palmitoyl-CoA ligase; Palmitoyl-CoA ligase 1; palmitoyl-CoA ligase 2; paltimoyl-CoA ligase 1; Phytanate--CoA ligase
Rabbit
Protein A
RUO
14081, 25288
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.