missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACTH Mouse anti-Human, Mouse, Rat, Clone: 3C12C7, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66358-1-IG-20UL
This item is not returnable.
View return policy
Description
ATCH (adrenocorticotropic hormone) is a hormone which plays a major role in stimulating the adrenal cortex. It is formed through cleavage of the polypeptide precursor proopiomelanocortin (POMC), which also results in several other cleavage products including MSH, ACTH, and beta endorphin. ATCH is secreted from the anterior pituitary in response to the corticotropin-releasing hormone from the hypothalamus. It stimulates the secretion of glucocorticoids like cortisol, but has little control over the stimulation of mineralocorticoids like aldosterone, which is another major hormone of the adrenal cortex.Specifications
| ACTH | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P01189, P01193, P01194 | |
| Pomc | |
| ACTH Fusion Protein Ag17311 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG1 |
| Immunofluorescence, Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| 3C12C7 | |
| Unconjugated | |
| POMC | |
| ACTH, ACTH Melanotropin alpha, Adrenocorticotropic hormone, Beta LPH Lipotropin gamma, CLIP, CLIP Lipotropin beta, Corticotropin lipotropin, Gamma LPH Melanotropin beta, LPH, MSH, POC, POMC, Pro opiomelanocortin, proopiomelanocortin | |
| Mouse | |
| Protein G | |
| RUO | |
| 18976, 24664, 5443 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction