Learn More
Erlab Activated Carbon Filtration At Extraction - Solvents - Solis 1200
Product Code. 18553756 Shop All Erlab Products
Change view
Click to view available options
Product Type:
Filter
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
18553756 Filter Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18553756 Supplier Erlab Supplier No. NORT000350/AS
  • ONLINE EXCLUSIVE PRICE
    1830.00€
    1697.00€ / Pack of 1
    Save 133.00€
    7% Off
 Restricted Item
Estimated Shipment: 12-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Filter for your Biological Safety Cabinet.
  • Activated carbon filtration at extraction for your BSC
  • Type of chemicals: Solvents
  • Compatible model: BSC Solis 1200
Specifications
Product Type Filter