Learn More
Activin C/Inhibin beta C Antibody (CL6915) [Janelia Fluor™ 646], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-76513JF646
Product Code. 30293948
  • 737.00€ / 0.10mL
Description
Activin C/Inhibin beta C Monoclonal antibody specifically detects Activin C/Inhibin beta C in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Activin C/Inhibin beta C
Monoclonal
Janelia Fluor 646
inhibin beta C chain, IHBC, inhibin, beta C
This antibody was developed using a recombinant protein derived from P55103, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cytokine Research
3626
Store at 4⪚C in the dark.
IgG1
Western Blot
CL6915
50mM Sodium Borate
Mouse
Protein A purified