Learn More
ACTR3 Monoclonal Antibody (OTI5G4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA812199
Description
The specific function of this gene has not yet been determined; however, the protein it encodes is known to be a major constituent of the ARP2/3 complex. This complex is located at the cell surface and is essential to cell shape and motility through lamellipodial actin assembly and protrusion.Specifications
| ACTR3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P61158 | |
| ACTR3 | |
| Human recombit protein fragment corresponding to amino acids 2-418 of human ACTR3 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI5G4 | |
| Unconjugated | |
| ACTR3 | |
| ARP3 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 10096 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.