Learn More
Socorex™ Acura™ Manual 825 Micropipette Triopack
Product Code. 10227032
Change view
Click to view available options
Volume (Metric):
0.5-10 μL, 10-100 μL, 100-1000 μL
10, 100, 1000 μL
10, 50, 200 μL
10-100 μL, 100-1000 μL, 0.5-5.0 mL
100-1000 μL, 0.5-5.0, 1.0-10 mL
2, 10, 50 μL
2, 20, 200 μL
2-20 μL, 20-200 μL, 100-1000 μL
5-50 μL, 20-200 μL, 100-1000 μL
Unit Size:
1 set
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
10227032 0.5-10 μL, 10-100 μL, 100-1000 μL 1 set
10680833 10, 100, 1000 μL 1 set
10112412 10, 50, 200 μL 1 set
10690833 2-20 μL, 20-200 μL, 100-1000 μL 1 set
10462812 5-50 μL, 20-200 μL, 100-1000 μL 1 set
10315622 10-100 μL, 100-1000 μL, 0.5-5.0 mL 1 set
10254312 100-1000 μL, 0.5-5.0, 1.0-10 mL 1 set
10142362 2, 10, 50 μL 1 set
10581003 2, 20, 200 μL 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 10227032 Supplier Socorex™ Supplier No. 825.TRIO.O
  • ONLINE EXCLUSIVE PRICE
    840.00€
    788.55€ / 1 sets
    Save 51.45€
    6% Off
 Restricted Item
Estimated Shipment: 15-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Classic and universal, these micropipettes are intended for all applications and come in a select three-pack. Socorex™ Acura™ Manual 825 Micropipette Triopack micropipettes are designed for comfort and sturdiness.
  • Ergonomic shape.
  • Lightweight.
  • Uultra-soft plunger stroke.
  • Smart and reliable volume adjustment:
    • State-of-the-art micrometric setting is performed smoothly and precisely by turning push button.
    • Finely machined click-stops and free rotating cap prevent any unwanted volume alteration.
  • Digital display of set volume on the front of the instrument is easily visible during pipetting.
  • Autoclavable.
Specifications
Certifications/Compliance EN ISO 8655, DIN 12650, GMP, GLP, GBEA, NCCLS and CE IVD Directive 98/79 EEC on medical devices
Autoclavable Autoclavable
Product Line Acura
Type Micropipette Triopack
Volume (Metric) 0.5-10 μL, 10-100 μL, 100-1000 μL
Includes 3 Acura™ micropipettes, manual, spike samples, Qualitips™ 10/100/1000μL
Quantity 3/Pk.