Learn More
ACVR1 Mouse anti-Human, Unconjugated, Clone: 1F11B10, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67417-1-IG-150UL
Description
The ACVR1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ACVR1. This antibody recognizes human, pig, mouse, rat antigen. The ACVR1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
| ACVR1 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat, Pig |
| Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
| 1F11B10 | |
| Unconjugated | |
| 150 μL | |
| 90 | |
| IgG2a |