Learn More
ACVR1 Mouse anti-Human, Unconjugated, Clone: 1F11B10, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67417-1-IG-20UL
 View more versions of this product
Product Code. 16840036
  • 145.00€ / 20µL
Description
The ACVR1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ACVR1. This antibody recognizes human, pig, mouse, rat antigen. The ACVR1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
ACVR1
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Mouse, Rat, Pig
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA
1F11B10
Unconjugated
20 μL
90