missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ACVR1 Polyclonal Antibody

Sheep Polyclonal Antibody
Brand: Invitrogen™ PA112641
This item is not returnable.
View return policy
Description
ACVR1 Polyclonal Antibody for Western Blot
Activin A receptor, type I (ACVR1) belongs to the transforming growth factor-beta (TGF-beta) superfamily of structurally related signaling proteins. ACVR1 signals a specific transcriptional response in concert with activin type II receptors. The ACVR1 gene encodes activin A type I receptor, which signals a particular transcriptional response in concert with activin type II receptors. Mutations in ACVR1 have been linked to Fibrodysplasia Ossificans Progressiva (FOP), a rare genetic disorder characterized by the formation of bone in muscles, tendons, and other connective tissues, as well as Epicanthus.
Specifications
| ACVR1 | |
| Polyclonal | |
| PBS with 0.08% sodium azide | |
| Q90ZK6 | |
| ACVR1 | |
| Recombinant protein corresponding to the extracellular domain of chicken ALK2. | |
| 250 μg | |
| Primary | |
| Chicken | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| ACVR1 | |
| activin A receptor type 1; activin A receptor type I; activin A receptor, type 1; activin A receptor, type I; activin A receptor, type II-like kinase 2; activin receptor I precursor; activin receptor type I; Activin receptor type-1; activin receptor-like kinase 2; activin type I receptor; ACTRI; ActR-I; ActRIA; Acvr; ACVR1; ACVR1A; ACVRLK2; ALK2; Alk-2; ALK2-B; Alk8; ATVR1; D330013D15Rik; FOP; hydroxyalkyl-protein kinase; serine/threonine-protein kinase receptor; serine/threonine-protein kinase receptor R1; SKR1; TGF-B superfamily receptor type I; Tgfb1; Tsk7L; TSK-7L; TSRI; TSR-I; type I TGF B receptor | |
| Sheep | |
| Ammonium Sulfate Precipitation | |
| RUO | |
| 395246 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction