Learn More
ACY3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA561683
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: MCSLPVPQEP LRRVAVTGGT HGNEMSGVYL ARHWLHAPAE LQRASFSAVP VLANPAATSG CRRYVDHDLN RTFTSSFLNS R Highest antigen sequence identity to the following orthologs: Mouse - 72%, Rat - 75%.
ACY3 belongs to the aspA/astE family. Aspartoacylase subfamily. Catalytic activity: N-acyl-L-aspartate + H(2)O = a carboxylate + L-aspartate.Specifications
ACY3 | |
Polyclonal | |
Unconjugated | |
Acy3 | |
0610006H10Rik; AA3; AAIII; Acy3; ACY-3; Acylase III; aminoacylase 3; Aminoacylase III; Aminoacylase-3; Aspa2; aspartoacylase (aminoacylase) 3; aspartoacylase (aminocyclase) 3; aspartoacylase 3; aspartoacylase-2; aspartoacylase-3; AW107362; EGK_05921; HCBP1; HCV core-binding protein 1; hepatitis C virus core-binding protein 1; LOW QUALITY PROTEIN: N-acyl-aromatic-L-amino acid amidohydrolase (carboxylate-forming); N-acyl-aromatic-L-amino acid amidohydrolase (carboxylate-forming); RGD1305394 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
91703 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96HD9 | |
Acy3 | |
Recombinant protein corresponding to Human HCBP1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only