missing translation for 'onlineSavingsMsg'
Learn More

Acyclovir Acetate, TRC

Product Code. 30011448
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011448 25 mg 25mg
30009591 50 mg 50mg
17953655 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011448 Supplier Toronto Research Chemicals Supplier No. TRCA19241025MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102728-64-3
Molecular Formula C10 H13 N5 O4
Molecular Weight (g/mol) 267.24
Synonym 2-[(2-Amino-6-oxo-1,6-dihydro-9H-purin-9-yl)methoxy]ethyl Acetate, Valaciclovir Hydrochloride Anhydrous Imp. I (EP), Aciclovir Imp. A (EP), Aciclovir Acetate, Acyclovir Acetate
IUPAC Name 2-[(2-amino-6-oxo-1H-purin-9-yl)methoxy]ethyl acetate
SMILES CC(=O)OCCOCn1cnc2C(=O)NC(=Nc12)N

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C10H13N5O4/c1-6(16)19-3-2-18-5-15-4-12-7-8(15)13-10(11)14-9(7)17/h4H,2-3,5H2,1H3,(H3,11,13,14,17)
Formula Weight 267.0968
Percent Purity >95%
Chemical Name or Material 2-[(2-Amino-6-oxo-1,6-dihydro-9H-purin-9-yl)methoxy]ethyl Acetate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.