missing translation for 'onlineSavingsMsg'
Learn More

Acyclovir Monophosphate, TRC

Product Code. 30240938
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30240938 10 mg 10mg
30240758 5 mg 5mg
30240552 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30240938 Supplier Toronto Research Chemicals Supplier No. TRCA19250010MG
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 66341-16-0
Molecular Formula C8 H12 N5 O6 P
Molecular Weight (g/mol) 305.18
IUPAC Name 2-[(2-amino-6-oxo-1H-purin-9-yl)methoxy]ethyl dihydrogen phosphate
SMILES NC1=Nc2c(ncn2COCCOP(=O)(O)O)C(=O)N1

Especificaciones

Quantity 10 mg
InChI Formula InChI=1S/C8H12N5O6P/c9-8-11-6-5(7(14)12-8)10-3-13(6)4-18-1-2-19-20(15,16)17/h3H,1-2,4H2,(H2,15,16,17)(H3,9,11,12,14)
Formula Weight 305.0525
Chemical Name or Material Acyclovir Monophosphate
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.