missing translation for 'onlineSavingsMsg'
Learn More

Acyclovir N,O-Diacetate, TRC

Product Code. 30135863
Change view
Click to view available options
Quantity:
250 mg
2.5 g
Unit Size:
2.5g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135863 2.5 g 2.5g
17961215 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30135863 Supplier Toronto Research Chemicals Supplier No. TRCA1679952.5G
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75128-73-3
Molecular Formula C12H15N5O5
Molecular Weight (g/mol) 309.28
Synonym 2-[[(2-(Acetylamino)-6-oxo-1,6-dihydro-9H-purin-9-yl]-methoxy]ethyl Acetate, Aciclovir Imp. G (EP)
IUPAC Name 2-[(2-acetamido-6-oxo-1H-purin-9-yl)methoxy]ethyl acetate
SMILES CC(=O)NC1=Nc2c(ncn2COCCOC(=O)C)C(=O)N1

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C12H15N5O5/c1-7(18)14-12-15-10-9(11(20)16-12)13-5-17(10)6-21-3-4-22-8(2)19/h5H,3-4,6H2,1-2H3,(H2,14,15,16,18,20)
Formula Weight 309.107
Chemical Name or Material 2-[((2-(Acetylamino)-6-oxo-1,6-dihydro-9H-purin-9-yl)-methoxy]ethyl Acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.