missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADAD2 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
486.00€ - 741.00€
Specifications
| Antigen | ADAD2 |
|---|---|
| Dilution | Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18304546
|
Novus Biologicals
NBP3-17119-25UL |
25 μg |
486.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18379095
|
Novus Biologicals
NBP3-17119-100UL |
100 μg |
741.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ADAD2 Polyclonal antibody specifically detects ADAD2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| ADAD2 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| adenosine deaminase domain containing 2, adenosine deaminase domain-containing protein 2, TENRLFLJ00337, Testis nuclear RNA-binding protein-like | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: CFPFSVSAELDGVVCPAGTANSKTEAKQQAALSALCYIRSQLENPESPQTSSRPPLAPLSVENILTHEQRCAALVSAGFDLLLDERSPYWACKGTVAGVILE | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS, pH 7.2, 40% glycerol | |
| 161931 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title