missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADAM10 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76973-0.025mg
This item is not returnable.
View return policy
Description
ADAM10 Polyclonal specifically detects ADAM10 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Frozen, Immunofluorescence, Immunocytochemistry.
Specifications
| ADAM10 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| a disintegrin and metalloprotease domain 10, a disintegrin and metalloproteinase domain 10, AD10, ADAM 10, ADAM metallopeptidase domain 10, CD156c, CD156c antigen, CDw156, disintegrin and metalloproteinase domain-containing protein 10, EC 3.4.24, HsT18717, kuz, Kuzbanian protein homolog, MADMEC 3.4.24.81, Mammalian disintegrin-metalloprotease | |
| Rabbit | |
| 85 kDa | |
| 0.025 mg | |
| Apoptosis, Signal Transduction | |
| 102 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 2 μg/mL, Immunocytochemistry/Immunofluorescence 10 μg/mL, Immunohistochemistry-Frozen | |
| NP_001101 | |
| ADAM10 | |
| Antibody was raised against a 17 amino acid peptide near the carboxy terminus of human ADAM10. The immunogen is located within the last 50 amino acids of ADAM10. Amino Acid Squence: PQRQRPRESYQMGHMRR | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction