missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADAM20 Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98032C
This item is not returnable.
View return policy
Description
ADAM20 Polyclonal specifically detects ADAM20 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| ADAM20 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| a disintegrin and metalloproteinase domain 20, ADAM 20, ADAM metallopeptidase domain 20, disintegrin and metalloproteinase domain-containing protein 20, EC 3.4.24, EC 3.4.24.-, EC 3.4.24.58 | |
| Produced in rabbits immunized with E. coli-derived Human ADAM20 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| DyLight 650 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 8748 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction