missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADAM28 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22234-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the ADAM (a disintegrin and metalloprotease domain) family. Members of this family are membrane-anchored proteins structurally related to snake venom disintegrins, and have been implicated in a variety of biological processes involving cell-cell and cell-matrix interactions, including fertilization, muscle development, and neurogenesis. The protein encoded by this gene is a lymphocyte-expressed ADAM protein. Alternative splicing results in two transcript variants. The shorter version encodes a secreted isoform, while the longer version encodes a transmembrane isoform.Specifications
| ADAM28 | |
| Polyclonal | |
| Unconjugated | |
| ADAM28 | |
| ADAM 28, ADAM28, eMDC II, eMDCII, MDC L, MDC Lm, MDC Ls, MDCL | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10863, 13522 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.34 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9JLN6, Q9UKQ2 | |
| ADAM28 | |
| ADAM28 Fusion Protein Ag17586 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction