Learn More
Fisherbrand™ Adapters for Sample Tubes
Product Code. 10732711
Change view
Click to view available options
For Use With (Equipment):
0.4mL sample tubes
0.5mL sample tubes
Unit Size:
Pack of 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
10732711 0.5mL sample tubes Pack of 6
11833503 0.4mL sample tubes Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10732711 Supplier Fisherbrand™ Supplier No. C1205
  • ONLINE EXCLUSIVE PRICE
    48.62€
    37.59€ / Pack of 6
    Save 11.03€
    22% Off
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
Excellent for use with micro centrifuges. Fisherbrand™ Adapters for Sample Tubes are durable and resilient.
  • White in color
  • Available for different tube sizes
Specifications
Type Adapter
For Use With (Equipment) 0.5mL sample tubes
No. per Pack 6/Pk.