missing translation for 'onlineSavingsMsg'
Learn More

Adenine, Serva Electrophoresis™

Product Code. 11777106
Encompass
Change view
Click to view available options
Quantity:
0.1 kg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11777106 0.1 kg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11777106 Supplier Serva Electrophoresis Supplier No. 1073903

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 73-24-5
Molecular Formula C5H5N5
MDL Number MFCD00041790
Quantity 0.1 kg
Synonym adenine, 6-aminopurine, 1h-purin-6-amine, 9h-purin-6-amine, vitamin b4, adenin, adeninimine, leuco-4, 6-amino-1h-purine, 6-amino-3h-purine
InChI Key GFFGJBXGBJISGV-UHFFFAOYSA-N
SMILES NC1=C2NC=NC2=NC=N1
IUPAC Name 7H-purin-6-amine
Molecular Weight (g/mol) 135.13
PubChem CID 190
ChEBI CHEBI:16708
Show More Show Less
compliance-icons
Product Identifier
  • Adenine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P330-Rinse mouth.
  • P405-Store locked up.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.