missing translation for 'onlineSavingsMsg'
Learn More

Adenine Sulfate Dihydrate, Hemisulfate, 99.66%, For HPLC analysis, MP Biomedicals™

Product Code. 11405032 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11405032 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11405032 Supplier MP Biomedicals Supplier No. 0219460790
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 6509-19-9
Color White
Assay Percent Range 0.9966
Molecular Formula C10H16N10O6S
MDL Number MFCD00150149
Quantity 500 g
Synonym 9h-purin-6-amine hemisulfate dihydrate, adenine hemisulfate dihydrate, 6-aminopurine sulfate dihydrate, bis ade sulfuric acid dihydrate, sulfuric acid bis vitamin b4 dihydrate, bis adenine sulfuric acid dihydrate, sulfuric acid-7h-purin-6-amine-water 1/2/2
InChI Key MKPCNMXYTMQZBE-UHFFFAOYSA-N
SMILES O.O.OS(O)(=O)=O.NC1=C2NC=NC2=NC=N1.NC1=C2NC=NC2=NC=N1
IUPAC Name bis(7H-purin-6-amine) sulfuric acid dihydrate
Molecular Weight (g/mol) 404.36
PubChem CID 44134512
Formula Weight 404.36
Physical Form Powder
Chemical Name or Material Adenine Sulfate Dihydrate, Hemisulfate, For HPLC analysis
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.