missing translation for 'onlineSavingsMsg'
Learn More

Adenosine-5′-diphosphoglucose Disodium Salt, TRC

Product Code. 30133646
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133646 10 mg 10mg
30008476 5 mg 5mg
30005390 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133646 Supplier Toronto Research Chemicals Supplier No. TRCA28170510MG
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102129-65-7
Molecular Formula C16 H23 N5 O15 P2 . 2 Na
Molecular Weight (g/mol) 633.3
Synonym ADP-Glucose Sodium Salt, P'-β-D-Glucopyranosyl Ester Adenosine 5'-(Trihydrogen Diphosphate) Disodium Salt, ADP-Glucose Disodium, ADP-glc Disodium, ADPG Disodium, ADP-D-Glucose Disodium Salt;
IUPAC Name disodium;[[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxy-tetrahydrofuran-2-yl]methoxy-oxido-phosphoryl] [(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydropyran-2-yl] phosphate
SMILES [Na+].[Na+].C[C@@H]1O[C@H](OP(=O)([O-])OP(=O)([O-])OC[C@H]2O[C@H]([C@H](O)[C@@H]2O)n3cnc4C(=O)NC(=Nc34)N)[C@@H](O)[C@H](O)[C@@H]1O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C16H25N5O15P2.2Na/c1-4-7(22)9(24)11(26)15(33-4)35-38(30,31)36-37(28,29)32-2-5-8(23)10(25)14(34-5)21-3-18-6-12(21)19-16(17)20-13(6)27;;/h3-5,7-11,14-15,22-26H,2H2,1H3,(H,28,29)(H,30,31)(H3,17,19,20,27);;/q;2*+1/p-2/t4-,5+,7+,8+,9+,10+,11-,14+,15+;;/m0../s1
Formula Weight 633.0461
Chemical Name or Material Adenosine-5'-diphosphoglucose Disodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.