Learn More
ADH4 Mouse anti-Human, Unconjugated, Clone: 3D12E9, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67303-1-IG-20UL
Description
The ADH4 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ADH4. This antibody recognizes human antigen. The ADH4 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| ADH4 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human |
| Western Blot, ELISA | |
| 3D12E9 | |
| Unconjugated | |
| 20 μL | |
| 127 | |
| IgG1 |