missing translation for 'onlineSavingsMsg'
Learn More

Adipoyl Chloride, TRC

Product Code. 30323340
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30323340 5 g 5g
30323328 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30323340 Supplier Toronto Research Chemicals Supplier No. TRCA2917005G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Adipoyl Chloride
CAS 111-50-2
Molecular Formula C6 H8 Cl2 O2
Quantity 5 g
Synonym Hexanedioyl Dichloride, 1,4-Bis(chlorocarbonyl)butane, Adipic Acid Dichloride, Adipoyl Dichloride, Adipyl Chloride, Adipyl Dichloride, Hexanedioic Acid Dichloride, Hexanedioic Dichloride
InChI Formula InChI=1S/C6H8Cl2O2/c7-5(9)3-1-2-4-6(8)10/h1-4H2
SMILES ClC(=O)CCCCC(=O)Cl
IUPAC Name hexanedioyl dichloride
Molecular Weight (g/mol) 183.03
Formula Weight 181.9901
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.