Learn More
Invitrogen™ ADK Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA578731
Description
Human Stefin B shares 78.6 % amino acid (aa) sequence identity with both mouse and rat Stefin B. Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human placenta tissue, human PC-3 whole cell, human T47D whole cell, human U937 whole cell, human U251 whole cell, human HepG2 whole cell, rat liver tissue, rat kidney tissue, rat stomach tissue, rat PC-12 whole cell, mouse kidney tissue, mouse stomach tissue, mouse NIH/3T3 whole cell. IHC: human placenta tissue, mouse liver tissue, rat liver tissue. ICC/IF: PC-3 cell.
The protein encoded by this gene catalyzes the addition of glucose monomers to the growing glycogen molecule through the formation of alpha-1,4-glycoside linkages. Mutations in this gene are associated with muscle glycogen storage disease. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
| ADK | |
| Polyclonal | |
| Unconjugated | |
| ADK | |
| 2310026J05Rik; 5033405D03Rik; Adenosine 5'-phosphotransferase; adenosine kinase; ADK; AI255373; AI987814; AK; testicular tissue protein Li 14 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 11534, 132, 25368 | |
| -20°C | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 500 μg/mL | |
| PBS with 4mg trehalose and no preservative | |
| P55263, P55264, Q64640 | |
| ADK | |
| E. coli-derived human ADK recombinant protein (Position: K165-T351). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.