Learn More
ADORA2B Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00€ - 410.00€
Specifications
| Antigen | ADORA2B |
|---|---|
| Concentration | 600 μg/mL |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17251366
|
Proteintech
21071-1-AP-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17241366
|
Proteintech
21071-1-AP-150UL |
150 μL |
410.00€
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The ADORA2B antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human ADORA2B. This antibody recognizes human, mouse antigen. The ADORA2B antibody has been validated for the following applications: WB,ELISA analysis.
Specifications
| ADORA2B | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| 136 |
| 600 μg/mL | |
| Polyclonal | |
| Liquid | |
| Human, Mouse | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.