Learn More
Mecmesin™ Advanced Force Gauge
The Advanced Force Gauge (AFG) is the most versatile, fully-featured digital instrument.
Brand: Mecmesin™ 850420
Additional Details : Weight : 0.95000kg
Description
With ever-increasing demands for improved quality, durability and ease-of-use of all products, there is a growing need for more physical testing during the manufacturing processes. The Advanced Force Gauge (AFG) is a powerful instrument packed full of innovative features designed to suit a breadth of tension and compression tests.
The AFG offers enhanced test features and data analysis capabilities:
- Max and 1st Peak mode - identify critical load points from the test
- Fast data-acquisition speed - captures break-force readings accurately
- Audible and visual pass/fail alarms - alerts operators immediately when testing
- Overload warning with trend bar - to minimize accidental damage to loadcell
- In-built storage for up to 500 readings - test rapidly now and download results later
- Data transfer via RS232, digimatic or analog - output data to PC or printer for further analysis
- Connection capability to external load cells and torque sensors - becomes a versatile display for many different applications
Specifications
1000 N | |
201 mm | |
70 mm | |
650 g |