Learn More
Motic™ AE31E Trinocular Microscope
Product Code. 15363087
Change view
Click to view available options
Head Style:
Trinocular
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Head Style Unit Size
15363087 Trinocular Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: This product is under eco-contribution tax.
Product Code. 15363087 Supplier Motic™ Supplier No. 1100100200576
  • ONLINE EXCLUSIVE PRICE
    4270.00€
    3838.25€ / Pack of 1
    Save 431.75€
    10% Off
View all promotions
Estimated Shipment: 11-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Regulatory: This product is under eco-contribution tax.
Use this microscope for routine live cell inspection. With an easy and flexible set up, it is ideal for University, routine-clinical and lab environments. Motic™ AE31E Trinocular Microscopes offer scale research functions, such as fluorescence and an externally mounted and centrable 30W/6V halogen Koehler illumination.
  • Widefield high eyepoint eyepieces WFPL10X/22mm with diopter adjustment on both eyepieces, with rubber eyecups
  • Centering telescope
  • Side facing quintuple nosepiece
  • CCIS Plan Achromatic objective PL4X
  • CCIS Plan Achromatic Phase objective PL PH10X, LWD PL PH20X, LWD PL PH40X
  • Plain stage, glass and metal inserts
  • ELWD N.A. 0.30 condenser
  • Phase slider, centrable PH1, brightfield, PH3
  • Koehler illumination quartz halogen 6V/30W with external lamp house and intensity control
Specifications
Head Style Trinocular