Learn More
Eppendorf™ Aerosol-tight caps
Product Code. 15783399 Shop All Eppendorf Products
Change view
Click to view available options
For Use With (Equipment):
Rotor S-4xUniversal-Large
S-4x1000 plate/tube bucket and S-4x750 plate bucket
Unit Size:
Pack of 2
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
15783399 S-4x1000 plate/tube bucket and S-4x750 plate bucket Pack of 2
15728409 Rotor S-4xUniversal-Large Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15783399 Supplier Eppendorf™ Supplier No. 5895111009
Add to basket
Add to basket
Use these caps with S-4x1000 plate/tube bucket, S-4x750 plate bucket or S-4x Universal Large Rotor
Set of 2 pcs.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) S-4x1000 plate/tube bucket and S-4x750 plate bucket