missing translation for 'onlineSavingsMsg'
Learn More

AF10 Antibody (S07-2G2), Novus Biologicals™

Rabbit Monoclonal Antibody

421.00€ - 645.00€

Specifications

Antigen AF10
Clone S07-2G2
Concentration 0.3 mg/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18691745
View Documents
Novus Biologicals
NBP3-19560-100ul
100 μg
645.00€
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18601884
View Documents
Novus Biologicals
NBP3-19560-50ul
50 μg
421.00€
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

AF10 Monoclonal antibody specifically detects AF10 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications

Specifications

AF10
0.3 mg/mL
Monoclonal
Purified
RUO
50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
8028
IgG
Affinity purified
S07-2G2
Western Blot
Unconjugated
Rabbit
Human, Mouse, Rat
AF10MGC75086, ALL1-fused gene from chromosome 10 protein, DKFZp686E10210, myeloid/lymphoid or mixed-lineage leukemia (trithorax (Drosophila) homolog);translocated to, 10, myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila);translocated to, 10, protein AF-10, type I AF10 protein
Recombinant protein of human AF10 (Uniprot # P55197)
Primary
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.