Learn More
AFAP1 Monoclonal Antibody (OTI2G9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA802329
Description
The protein encoded by this gene is a Src binding partner. It may represent a potential modulator of actin filament integrity in response to cellular signals, and may function as an adaptor protein by linking Src family members and/or other signaling proteins to actin filaments. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
| AFAP1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8N556 | |
| AFAP1 | |
| Human recombit protein fragment corresponding to amino acids 100-386 of human AFAP1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2G9 | |
| Unconjugated | |
| AFAP1 | |
| AFAP, AFAP-110, AFAP110 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 60312 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.