missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Afatinib-d6 Dimaleate (N,N-dimethyl-d6), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | (2 Z)-Afatinib-d6 |
| Molecular Formula | C24 D6 H19 Cl F N5 O3 . 2 C4 H4 O4 |
| Quantity | 0.01 g |
| Synonym | (2 E)-N-[4-[(3-Chloro-4-fluorophenyl)amino]-7-[[(3 S)-tetrahydro-3-furanyl]oxy]-6-quinazolinyl]-4-[di(methyl-d3)amino]-2-butenamide (2 Z)-2-butenedioate (1:2), Afatinib-d6 dimaleate, Afatinib-D6 (N,N-dimethyl-D6) dimaleate, Afatinib-d6 Dimaleate (N,N-dimethyl-d6) |
| InChI Formula | InChI=1 S/C24H25ClFN5O3.2C4H4O4/c1-31(2)8-3-4-23(32)30-21-11-17-20(12-22(21)34-16-7-9-33-13-16)27-14-28-24(17)29-15-5-6-19(26)18(25)10-15;2*5-3(6)1-2-4(7)8/h3-6,10-12,14,16 H,7-9,13H2,1-2H3,(H,30,32)(H,27,28,29);2*1-2 H,(H,5,6)(H,7,8)/b4-3+;2*2-1-/t16-;;/m0../s1/i1D3,2D3;; |
| SMILES | [2 H]C([2 H])([2 H])N(C\C=C\C(=O)Nc1cc2c(Nc3ccc(F)c(Cl)c3)ncnc2cc1O[C@H]4 CCOC4)C([2 H])([2 H])[2 H].OC(=O)\C=C/C(=O)O.OC(=O)\C=C/C(=O)O |
| IUPAC Name | (E)-4-[bis(trideuteriomethyl)amino]-N-[4-(3-chloro-4-fluoroanilino)-7-[(3 S)-oxolan-3-yl]oxyquinazolin-6-yl]but-2-enamide;(Z)-but-2-enedioic acid |
| Molecular Weight (g/mol) | 724.1197 |
| Formula Weight | 723.2226 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction