Learn More
Aflatoxin G2, TRC
Specifications
Specifications
| Chemical Name or Material | Aflatoxin G2 |
| CAS | 7241-98-7 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C17 H14 O7 |
| Quantity | 1 mg |
| Synonym | 1H,12H-Furo[3',2':4,5]furo[2,3-h]pyrano[3,4-c][1]benzopyran-1,12-dione, 3,4,7a,9,10,10a-hexahydro-5-methoxy-, (7aR-cis)-, 1H,12H-Furo[3',2':4,5]furo[2,3-h]pyrano[3,4-c][1]benzopyran-1,12-dione, 3,4,7aα,9,10,10aα-hexahydro-5-methoxy- (8CI), Aflatoxin G2 (7CI), (7aR,10aS)-3,4,7a,9,10,10a-Hexahydro-5-methoxy-1H,12H-furo[3',2':4,5]furo[2,3-h]pyrano[3,4-c][1]benzopyran-1,12-dione, Dihydroaflatoxin G1 |
| InChI Formula | InChI=1S/C17H14O7/c1-20-9-6-10-12(8-3-5-22-17(8)23-10)14-11(9)7-2-4-21-15(18)13(7)16(19)24-14/h6,8,17H,2-5H2,1H3/t8-,17+/m0/s1 |
| SMILES | COc1cc2O[C@H]3OCC[C@H]3c2c4OC(=O)C5=C(CCOC5=O)c14 |
| IUPAC Name | (3S,7R)-11-methoxy-6,8,16,20-tetraoxapentacyclo[10.8.0.02,9.03,7.013,18]icosa-1,9,11,13(18)-tetraene-17,19-dione |
| Molecular Weight (g/mol) | 330.29 |
| Show More |