Learn More
LNI Swissgas AG OFCAS iQ 220 8
Product Code. 17882039 Shop All LNI Swissgas Products
Change view
Click to view available options
Voltage:
100/115/220/230 V
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Voltage unitSize
17882039 100/115/220/230 V Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: This product is under eco-contribution tax.
Product Code. 17882039 Supplier LNI Swissgas Supplier No. 6920.47.220
  • ONLINE EXCLUSIVE PRICE
    12970.00€
    11809.52€ / Pack of 1
    Save 1160.48€
    8% Off
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
Add to basket
Regulatory: This product is under eco-contribution tax.
AG OFCAS-Scroll with dryer integrated.
  • AG OFCAS-Scroll 220 L/min at 8 bar, 230 V with dryer integrated.
Specifications
Voltage 100/115/220/230 V
Type Air Generator