missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AGER Antibody (CL10587) [mFluor Violet 500 SE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07982MFV500
This item is not returnable.
View return policy
Description
AGER Monoclonal antibody specifically detects AGER in Human,Rat samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
Specifications
| AGER | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| advanced glycosylation end product-specific receptor, RAGE isoform delta, RAGE isoform sRAGE-delta, Receptor for advanced glycosylation end products, receptor for advanced glycosylation end-products | |
| This antibody was developed using a synthetic peptide derived from Q15109, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cancer | |
| 177 | |
| Store at 4⪚C in the dark. | |
| IgG2a |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CL10587 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction