missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aggrecan Antibody (Cat-315), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-29796
This item is not returnable.
View return policy
Description
Aggrecan Monoclonal specifically detects Aggrecan in Rat, Feline samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
Specifications
| Aggrecan | |
| Monoclonal | |
| Unconjugated | |
| Ascites with 0.1% Sodium Azide | |
| AGC1SEDK, aggrecan, aggrecan core protein, Cartilage-specific proteoglycan core protein, Chondroitin sulfate proteoglycan 1, Chondroitin sulfate proteoglycan core protein 1, CSPCP, CSPG1aggrecan 1, CSPGCP, large aggregating proteoglycan, MSK16AGCAN | |
| Mouse | |
| Unpurified | |
| RUO | |
| Primary | |
| Chondroitin sulfate proteoglycan (CSPG), protein core epitope. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgM |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| Cat-315 | |
| Western Blot 1:5000, Immunohistochemistry 1:2000, Immunocytochemistry/Immunofluorescence 1:500, Immunoprecipitation 1:10-1:500 | |
| P16112 | |
| ACAN | |
| Feline brain proteoglycans. | |
| 0.1 mL | |
| Extracellular Matrix, Growth and Development, Mesenchymal Stem Cell Markers, Neuronal Cell Markers, Neuronal Stem Cell Markers, Stem Cell Markers | |
| 176 | |
| Rat, Feline | |
| Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction