missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AGR3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584177
This item is not returnable.
View return policy
Description
Immunogen sequence: WGDDITWVQT YEEGLFYAQK SKKPLMV Highest antigen sequence indentity to the following orthologs: Mouse - 89%, Rat - 89%.
AGR3 (Anterior Gradient 3) protein, also known as AG3 (hAG-3, HAG3 in human), or BCMP11, is a secreted cytoplasmic protein which is involved in metastasis induction and p53 tumour supressor inhibition. It may serve as molecular marker and potential therapeutic target for hormone-responsive breast tumours. Its Xenopus homolog is associated with anteroposterior fate determination during early development.
Specifications
| AGR3 | |
| Polyclonal | |
| Unconjugated | |
| AGR3 | |
| AG3; AG-3; AGR3; anterior gradient 3; anterior gradient 3 homolog; anterior gradient 3, protein disulphide isomerase family member; anterior gradient homolog 3; anterior gradient protein 3; anterior gradient protein 3 homolog; BCMP11; Breast cancer membrane protein 11; E030025L21Rik; Gm888; H PDIA18; HAG3; hAG-3; PDIA18; Protein disulfide isomerase family A, member 18; RGD1565406; UNQ642/PRO1272 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 155465 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8TD06 | |
| AGR3 | |
| Recombinant protein corresponding to Human AGR3. Recombinant protein control fragment (Product #RP-101496). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction